Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sera-Mag™ Carboxylate-Modified Magnetic Beads and SpeedBeads

Catalog No. 09928213 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
09-928-213 1 mL
09-928-215 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-928-213 Supplier Cytiva Supplier No. 29729998
Add to Cart
Add to Cart

For covalent coupling of proteins and amine-modified ligands based on Sera-Mag™ and SpeedBead technology for fast magnetic separation.

Sera-Mag™ Carboxylate-Modified Magnetic Beads & SpeedBeads have free carboxyl groups on the surface for convenient covalent coupling of target molecules (proteins, peptides, and amine-modified ligands) via primary amines (NH2).

  • Wide application range: Carboxylic groups on the surface form covalent amide bonds with proteins, peptides or amine-modified ligands for use in many downstream applications including protein purification, DNA sample preparation and clean-up, protenomicsand immunoassays.
  • Convenient one-step or two-step coupling: Carbodiimide chemistry allows rapid formation of stable bonds to minimize ligand leaching.
  • Fast magnetic separation: Available in original Sera-Mag™ format or Sera-Mag™ SpeedBeads version for faster magnetic response and shorter assay times in clinical diagnostic tests.
  • High performance: Excellent sensitivity and low nonspecific binding for greater accuracy.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 50 mg/mL
Content And Storage 4°C
Contents Magnetic Beads and Speed Beads
Color Black
Material Magnetite
Product Type Dye Reagents
Ligand Type Carboxylated Polymer Surface
Volume (Metric) 1 mL
Diameter (Metric) 3 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.