Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sera-Mag™ Oligo d(T) Magnetic Beads

Catalog No. 09880012 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-880-012 1 mL
09-981-145 5 mL
09-880-014 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-880-012 Supplier Cytiva Supplier No. 38152103011150
Add to Cart
Add to Cart

Suited for capturing or isolating mRNA from a variety of sources. Due to their cauliflower like structure, Sera-Mag beads have high binding capacity. Oligo dT magnetic beads contain covalently bound oligo(dT)14.

  • Colloidally stable and will remain in suspension for extended periods of time in the absence of a magnetic field
  • High binding capacity
  • 1.0μm diameter
  • High oligo dT content for mRNA binding
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1% solids (10mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) mRNA Purification
Quantity 1 mL
Diameter (Metric) 1 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.