Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sera-Mag SpeedBeads™ Protein A/G Magnetic Beads

Catalog No. 09981920
Click to view available options
Quantity:
1 mL
5 mL
100 mL

Provide a fast, convenient method for manual and automated magnetic isolation of IgA and IgG proteins using affinity binding. The beads can be used with the broadest range of antibody species and subclasses (even when subclass identity has not been determined).

  • Their cauliflower-like structure gives Sera-Mag beads high binding capacity
  • Fast and easy IgA purification and IgG purification
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1% solids (10mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) Affinity Purification
Quantity 1 mL
Diameter (Metric) 1 μm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.