Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sera-Mag SpeedBeads™ Protein A/G Magnetic Beads

Catalog No. 09981921 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-981-921 5 mL
09-981-922 100 mL
09-981-920 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-981-921 Supplier Cytiva Supplier No. 17152104010150
Add to Cart
Add to Cart

Provide a fast, convenient method for manual and automated magnetic isolation of IgA and IgG proteins using affinity binding. The beads can be used with the broadest range of antibody species and subclasses (even when subclass identity has not been determined).

  • Their cauliflower-like structure gives Sera-Mag beads high binding capacity
  • Fast and easy IgA purification and IgG purification
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1% solids (10mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) Affinity Purification
Quantity 5 mL
Diameter (Metric) 1 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.