Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sera-Mag SpeedBeads Streptavidin-Blocked Magnetic Particles

Catalog No. 09981911 Shop All Cytiva Products
Change view
Click to view available options
Packaging:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
09-981-911 1 mL
09-981-912 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-981-911 Supplier Cytiva Supplier No. 21152104011150
Add to Cart
Add to Cart

Combines high affinity for biotinylated target molecules with very low nonspecific binding.

  • Combines fast reaction kinetics with very low non-specific binding for increased throughput and precision
  • Applications include automated immunoassays, immunoprecipitation, and protein purification
  • Streptavidin coating targets Biotinylated Molecules
  • Compounds that are difficult to attach to particle surfaces by conventional means may be amenable to biotinylation
  • Provides a high biotin-binding capacity along with a strong affinity for targeted, biotin labeled molecules
  • The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer
  • Two layers of magnetite are coated onto this core, resulting in faster magnetic response times
  • The surface is blocked with a non-surfactant, non-protein based method, to help prevent non-specific binding of proteins
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 10 mg/mL
    Packaging 1 mL
    Diameter (Metric) 1 μm
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.