Learn More
Cytiva Sera-Mag Streptavidin-Coated Magnetic Particles
GSA_VA
Catalog No. 45002359 Shop All Cytiva Products
Change view
Click to view available options
Binding Property:
2500 to 3500 pmol/mg
3500 to 4500 pmol/mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Binding Property
45-002-359 2500 to 3500 pmol/mg
45-002-360 3500 to 4500 pmol/mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-359 Supplier Cytiva Supplier No. 30152103010150
Add to Cart
Add to Cart
Exhibits high affinity for biotinylated target molecules. Cytiva Sera-Mag Streptavidin-Coated Magnetic Particles combine fast reaction kinetics and low non-specific binding for increased throughput and precision. Can be used as a universal base particle for coating ligands to the particle surface.
  • Ideal for immunoassay and molecular biology applications, including sample preparation and assay development for genomics and proteomics
  • Available with various nominal biotin binding capacities for optimizing assay development
  • Low dissociation constant to tightly bound ligands
  • Low non-specific binding for greater precision and accuracy
  • Even streptavidin coating for reliable results
  • Uniform, nominal 1μm diameter provides high surface area and excellent lot-to-lot reproducibility
Streptavidin Coating Targets Biotinylated Molecules
Compounds that are difficult to attach to particle surfaces by conventional means may be amenable to biotinylation. Sera-Mag Streptavidin-Coated Magnetic Particles provide a high biotin-binding capacity along with a strong affinity for targeted, biotin labeled molecules.Fast Magnetic Response Times and Stable Physical Integrity
The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer. A layer of magnetite is then coated onto this core.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Binding Property 2500 to 3500 pmol/mg
Content And Storage 4°C
Quantity 5 mL
Diameter (Metric) 1 μm