Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sera-Mag™ Streptavidin Magnetic Bead, medium biotin binding capacity

Catalog No. 09981129 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-981-129 1 mL
09-981-131 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-981-129 Supplier Cytiva Supplier No. 30152104011150
Add to Cart
Add to Cart

Their cauliflower-like structure gives Sera-Mag beads high binding capacity. Sera-Mag Streptavidin beads contain covalently bound streptavidin and are available with low, medium, or high nominal biotin binding capacities. Choose the biotin-binding capacity needed to optimize your application.

Biotin Binding Capacity of 3500 to 4500pmol/mg beads
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1% solids (10mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets
Quantity 1 mL
Diameter (Metric) 1 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.