Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERAC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | SERAC1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SERAC1 Polyclonal specifically detects SERAC1 in Human samples. It is validated for Western Blot.Specifications
SERAC1 | |
Polyclonal | |
Rabbit | |
Cancer | |
FLJ14917, FLJ30544, protein SERAC1, serine active site containing 1 | |
SERAC1 | |
IgG | |
74 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_116250 | |
84947 | |
Synthetic peptide directed towards the N terminal of human SERAC1The immunogen for this antibody is SERAC1. Peptide sequence KTLIGLARSEESDLRFFLLPPPLPSLKEDSSTEEELRQLLASLPQTELDE. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title