Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERAC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545141
Description
Peptide sequence: KTLIGLARSE ESDLRFFLLP PPLPSLKEDS STEEELRQLL ASLPQTELDE Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 85%; Horse: 79%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 93%; Rat: 100%.
The function of this protein remains unknown.
Specifications
| SERAC1 | |
| Polyclonal | |
| Unconjugated | |
| Serac1 | |
| 4930511N22Rik; D17Ertd141e; hypothetical protein LOC519860; protein SERAC1; Protein SERAC1-like protein; RGD1565064; Serac1; serine active site containing 1; serine active site-containing protein 1; srac1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84947 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96JX3 | |
| Serac1 | |
| Synthetic peptide directed towards the N-terminal of human SERAC1 (aa 176-225). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction