Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERAC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555244
Description
Immunogen sequence: KPMEDEDRYT TCWPKTWLAK DCPALRIISV EYDTSLSDWR ARCPMERKSI AFRSNELLRK LRAAGVGDRP VVWISHSMGG LLVKKMLLEA STKPEMS Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
The function of this protein remains unknown.
Specifications
| SERAC1 | |
| Polyclonal | |
| Unconjugated | |
| Serac1 | |
| 4930511N22Rik; D17Ertd141e; hypothetical protein LOC519860; protein SERAC1; Protein SERAC1-like protein; RGD1565064; Serac1; serine active site containing 1; serine active site-containing protein 1; srac1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84947 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96JX3 | |
| Serac1 | |
| Recombinant protein corresponding to Human SERAC1. Recombinant protein control fragment (Product #RP-94734). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction