Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERAC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555245
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555245 Supplier Invitrogen™ Supplier No. PA555245
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RVIGNMALNE HLHSSIVRSG WVSIMAEAMK SPHIMESSHA ARILANLDRE TVQEKYQDGV YVLHPQYRTS QPIKA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERAC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Serac1
Gene Accession No. Q96JX3
Gene Alias 4930511N22Rik; D17Ertd141e; hypothetical protein LOC519860; protein SERAC1; Protein SERAC1-like protein; RGD1565064; Serac1; serine active site containing 1; serine active site-containing protein 1; srac1
Gene Symbols Serac1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SERAC1. Recombinant protein control fragment (Product #RP-94735).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84947
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.