Learn More
SeraCare™ ACCURUN™ 644 Cystic Fibrosis 40plus4 Control
Shop All SeraCare Life Sciences Products
Click to view available options
Quantity:
100 μL
Description
- Identifies mutations in the cystic fibrosis transmembrane conductance regulator (CFTR) gene
- Covers all 47 mutations in the Prima xTAG™ CFTR IVD Assay
- Eliminate errors arising from use of rotating gDNA control schemes with incomplete mutation profiles
- Controls contain a mixture of oligonucleotides that produce a call for every mutation tested
- All mutations contained in a single vial
- High purity, large lots, stable materials–no additional processing required
Not for use in diagnostic procedures
Specifications
Specifications
Certifications/Compliance | CE, RUO |
Type | ACCURUN 644 Cystic Fibrosis 40+4 Control |
Intended Use | Detects significant errors with the use of rotating gDNA control schemes, research use only |
Quantity | 100 μL |