Learn More
SeraCare™ ACCURUN™ 676 Cystic Fibrosis Extended Control
Catalog No. 22554480 Shop All SeraCare Life Sciences Products
Click to view available options
Quantity:
100 μL
Catalog No. 22-554-480
Supplier: SeraCare™ 20400009
Only null left
Add to Cart
Add to Cart
A synthetic, multiplex control formulated for research use in laboratory testing, molecular screening, and mutation testing, with assays that identify mutations in the cystic fibrosis transmembrane conductance regulator (CFTR) gene
  • Eliminates errors arising from use of rotating gDNA control schemes with incomplete mutation profiles
  • Controls contain a mixture of oligonucleotides that produce a call for every mutation tested
  • ACCURUN 676 covers 34 mutations, and when used in conjunction with ACCURUN 644, covers all 81 mutations in the Prima xTAG™ CFTR 70+6 Assay
  • All mutations contained in a single vial
  • High purity, large lots, stable materials–no additional processing required
Not for use in diagnostic procedures
Specifications
Certifications/Compliance RUO
Type ACCURUN 676 Cystic Fibrosis Extended Control
Intended Use Detects significant errors with the use of rotating gDNA control schemes, research use only
Quantity 100 μL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.