Learn More
Description
- Combine LabLink xL with the ACCURUN products for an IVD exclusive infectious disease and nucleic acid testing Quality Assurance Program (QAP) from Fisher HealthCare
- With the LabLink xL QAP program, a lab's performance can be compared to peer laboratories' performances and used to benchmark performance to group standards:
- Track how other lab's signal to cutoff ratio (s/co) or recovered values (mean values) compare
- Compare how a lab's variability (standard deviation) compares to a group average
- Determine if a lab's s/co or recovered values are statistically within acceptable group limits
Specifications
Specifications
| Certifications/Compliance | IVD, CE, HC |
| Content And Storage | 2°C to 8°C |
| Type | ACCURUN 24 Multi-analyte Confirmatory Control |
| Intended Use | Detection of Antibodies to HIV 1 and 2, HTLV I, Hepatitis C Virus |
| Matrix | Human Serum |
| Quantity | 3 x 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.