Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SeraCare™ AccuSpan™ Linearity Panels
Shop All SeraCare Life Sciences Products
Click to view available options
Type:
Epstein Barr Virus (EBV)
Herpes Simplex Virus 1 (HSV1)
Herpes Simplex Virus 2 (HSV2)
JC Virus (JCV)
Varicella Zoster Virus (VZV)
Description
- Each panel consists of four members: one negative, one low positive, one medium positive and one high positive sample
- Comes in ready-to-go, single-use format with target concentrations that cover range of most molecular assays
- Full size: 4 x 1mL
Specifications
Specifications
Type | Varicella Zoster Virus (VZV) |
Includes | 1 negative, 1 low positive, 1 medium positive, 1 high positive sample |
Quantity | 4 x 1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction