Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ Anti-HCV Qualification Panel

Catalog No. 22554562 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
6 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-562 6 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Members of the Anti-HCV Qualification Panel are manufactured from human serum or plasma that is negative for HBsAg and antibodies to HIV 1 and 2, and HTLV. Anti-HCV positive stock materials are treated with beta-propiolactone and ultraviolet radiation. This does not ensure the absence of pathogens, and these materials should be handled following good laboratory safety practice. Do not pipette by mouth. Do not smoke, eat or drink in areas where specimens are handled. Dispose of this product in biohazard containers.
Catalog No. 22-554-562 Supplier SeraCare™ Supplier No. 24000148
Add to Cart
Add to Cart
Members of the Anti-HCV Qualification Panel are manufactured from human serum or plasma that is negative for HBsAg and antibodies to HIV 1 and 2, and HTLV. Anti-HCV positive stock materials are treated with beta-propiolactone and ultraviolet radiation. This does not ensure the absence of pathogens, and these materials should be handled following good laboratory safety practice. Do not pipette by mouth. Do not smoke, eat or drink in areas where specimens are handled. Dispose of this product in biohazard containers.

Panel for training, qualifying and re-qualifying in the performance of tests for the detection of anti-HCV

The Anti-HCV Qualification Panel is a panel of six members with established reactivity in anti-HCV (Hepatitis C Virus) assays; may be used for training, qualifying and re-qualifying technical personnel

  • May also be used as part of ongoing programs of lot acceptance and internal proficiency testing for anti-HCV assays, to isolate system errors and in troubleshooting these assays, as a component of a quality assurance program

  • Extensive array of panels helps labs maintain the highest standards for assay validation, regulatory compliance and method development
  • Each panel includes comprehensive, comparative data from a variety of methods and commercially available test kits, allowing reagent, assay and platform performance, sensitivity and specificity to be independently challenged
  • Assembled from highly characterized, disease state patient samples, and developed specifically for infectious disease testing
  • Enables labs to evaluate, monitor and troubleshoot test and staff performance
  • Available for most instrument platforms and test methods
  • Minimizes down time and accelerates verification of new test methods and platforms

  • Rigorously tested and characterized using highest-quality test methods
  • Comprehensive test results provided with each panel; quantified against international standards when available
  • Long-term stability and availability
  • All SeraCare facilities are ISO 9001 and 13485 certified and comply with cGMP standards

Specifications

Certifications/Compliance ISO 9000 13485
Content And Storage 2°C to 8°C
Format Panel
Type Anti-HCV Qualification Panel Level 2
Includes Set of six members: five panel members formulated with various reactivities for Anti-HCV, nonreactive member formulated from Anti-HCV nonreactive pools
Intended Use Used for training, qualifying, and re-qualifying technical personnel in the performance of tests for the detection of Anti-HCV
Matrix Human Serum or Plasma
Quantity 6 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.