Learn More
SeraCare™ Hepatitis B Seroconversion Panels
Shop All SeraCare Life Sciences Products
Click to view available options
Quantity:
6 x 1.5 mL
8 x 1.5 mL
Description
For research use only.
Specifications
Specifications
| Format | Panel |
| Type | Hepatitis B Seroconversion Panel |
| Intended Use | For evaluating HBV test system using well characterized specimens, to provide comprehensive data for comparative analysis |
| Quantity | 6 x 1.5 mL |