Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SeraCare™ Hepatitis C Seroconversion Panels
Click to view available options
Format:
Genotype 1a Panel
Genotype 3a Panel
Description
- Panel helps labs maintain the highest standards for assay validation, regulatory compliance and method development
- Each panel includes comprehensive, comparative data from a variety of methods and commercially available test kits, allowing reagent, assay/platform performance, sensitivity and specificity to be independently challenged
- Assembled from highly characterized, disease state patient samples, and developed specifically for infectious disease testing, both serology and NAT
- Minimizes down time and accelerates verification of new test methods, reagent systems and platforms
- SeraCare panels are rigorously tested and characterized using the highest quality test methods
- Comprehensive test results provided with each panel; quantified against international standards when available
- Long-term stability and availability
- All SeraCare facilities are ISO 9001 and 13485 certified and comply with cGMP standards
HCV Seroconversion Panel Mfr. No. PHV922-1.2
- HCV genotype 3a
- Undiluted aliquots from plasma units collected from a 22-year old male in the US Midwest in 2008
- HCV genotype 1a
- Undiluted aliquots from plasma units collected from a 30-year old male in the U.S. Northeast in 2008
Specifications
Specifications
Content And Storage | -65°C to -80°C |
Format | Genotype 3a Panel |
Type | Hepatitis C Seroconversion Panel |
Intended Use | For evaluating HCV test system using well characterized specimens, to provide comprehensive data for comparative analysis |
Quantity | 6 x 1.2 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction