Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SeraCare™ HIV-1 Seroconversion Panels
Click to view available options
Quantity:
4 x 1.2 mL
5 x 1.2 mL
7 x 1.2 mL
Content And Storage:
-10°C
-65°C to -80°C
-70°C
Time Range:
11 Days in 2000
14 Days in 2002
15 Days in 2002
16 Days in 2000
33 Days in 2010
9 Days in 1999
Description
- Group of serial bleeds from an individual plasma donor during seroconversion
- Contains set of undiluted plasma samples from single plasma donor
- 1 vial per member, 1.2mL per vial
Cat. No. 21-123-109:
- Four member panel
- Collected over an 11-day period in 2000
- Member 3 is a low titer HIV antigen sample, challenging for antigen and Ag/Ab tests
- Member 4 is a challenging sample for antibody assays
- Four member panel
- Collected over an 16-day period in 2000
- PRB974-02 is a moderate positive HIV RNA sample, challenging for HIV antigen assays
- PRB974-03 is detected at very low signal to cutoff by all HIV Ag/Ab methods tested
- PRB974-04 is a very challenging sample for anti-HIV assays
- Five member panel
- Collected over an 14-day period in 2002
- PRB975-03 is a challenging sample for HIV RNA assays
- Demonstrates improvement in sensitivity with introduction of “4th generation” HIV Ag/Ab tests
- Four member panel
- Collected over an 9-day period in 1999
- Member 2 is a potentially challenging sample for HIV antigen and Ag/Ab tests
- Demonstrates improvement in sensitivity with introduction of “4th generation” HIV Ag/Ab tests
- Four member panel
- Collected over an 15-day period in 2002
- Member 1 is a challenging HIV RNA sample
- Members 3 and 4 are both challenging for some antibody assays
- Seven member panel
- Collected over an 33-day period in 2010
Specifications
Specifications
Content And Storage | -70°C |
Type | HIV-1 Seroconversion Panel |
Intended Use | Evaluating HIV-1 test system using well characterized specimens, to provide comprehensive data for comparative analysis |
Quantity | 7 x 1.2 mL |
Time Range | 33 Days in 2010 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction