Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ HIV-1 Seroconversion Panels AD - AZ

Catalog No. 22554466 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
4 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-466 4 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-554-466 Supplier SeraCare™ Supplier No. 06000230
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Undiluted samples collected from a single donor during development of infection and subsequent immune response

  • Includes comprehensive, comparative data from a variety of methods and commercially available test kits, allowing user to independently challenge reagent, assay/platform performance, sensitivity and specificity
  • Assembled from highly characterized, disease state patient samples, and developed specifically for infectious disease testing, both serology and NAT
  • Minimizes down time and accelerates verification of new test methods, reagent systems, and platforms

Specifications

Content And Storage -10°C
Format Panel
Type HIV-1 Seroconversion Panel
Includes Set of 4 undiluted Plasma samples from a single Plasma donor collected during a period of HIV-1 seroconversion
Intended Use Evaluating Anti-HIV, HIV-1 Antigen, and HIV-1 RNA assays with well characterized specimens, to provide comprehensive data for comparative analysis
Quantity 4 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.