Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ Seroconversion Panels

Catalog No. 22554467 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-554-467 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
These materials have not been inactivated and should be handled, like all blood products, as if potentially biohazardous, following good laboratory safety practice. Panel members have been tested and found negative for HBsAg, anti-HCV and anti-HIV-1/2. This does not ensure the absence of these or other human pathogens. Never pipette by mouth. Do not smoke, eat or drink in areas where specimens are handled. These materials should be disposed of in a manner that will inactivate pathogenic agents.
Catalog No. 22-554-467 Supplier SeraCare™ Supplier No. 06150016
Add to Cart
Add to Cart
These materials have not been inactivated and should be handled, like all blood products, as if potentially biohazardous, following good laboratory safety practice. Panel members have been tested and found negative for HBsAg, anti-HCV and anti-HIV-1/2. This does not ensure the absence of these or other human pathogens. Never pipette by mouth. Do not smoke, eat or drink in areas where specimens are handled. These materials should be disposed of in a manner that will inactivate pathogenic agents.

Panel of undiluted serial bleeds collected during the evolution of the donor's immune response to Hepatitis A infection

PHT903 is a panel of undiluted plasma samples collected from a single donor during development of Hepatitis A infection and subsequent immune response

  • Panel converts positive to negative for HAV RNA and negative to positive for anti-HAV IgM and anti-HAV Total
  • For research use only

  • Panel helps labs maintain the highest standards for assay validation, regulatory compliance and method development
  • Each panel includes comprehensive, comparative data from a variety of methods and commercially available test kits, allowing reagent, assay/platform performance, sensitivity and specificity to be independently challenged
  • Assembled from highly characterized, disease state patient samples, and developed specifically for infectious disease testing, both serology and NAT
  • Minimizes down time and accelerates verification of new test methods, reagent systems and platforms

  • SeraCare panels are rigorously tested and characterized using the highest quality test methods
  • Comprehensive test results provided with each panel; quantified against international standards when available
  • Long-term stability and availability
  • All SeraCare facilities are ISO 9001 and 13485 certified and comply with cGMP standards

Specifications

Certifications/Compliance ISO 9000 13485
Content And Storage -10°C
Format Panel
Type Hepatitis A Seroconversion Panel
Intended Use For evaluating assays for markers of Hepatitis A infection, to study the Humoral immune response to this infection
Quantity 10 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.