Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva SeraSil-Mag Silica Coated Superparamagnetic Beads

Catalog No. 09928103 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 mL
60 mL
Size:
400 nm diameter particle
400 nm diameter particles
700 nm diameter particle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Size
09-928-103 60 mL 400 nm diameter particle
09-928-102 5 mL 400 nm diameter particles
09-928-104 5 mL 700 nm diameter particle
09-928-105 60 mL 700 nm diameter particle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 09-928-103 Supplier Cytiva Supplier No. 29357371
Add to Cart
Add to Cart
This item is not returnable. View return policy

SeraSil-Mag beads for nucleic acid isolation deliver high purity DNA extraction in NGS applications.

  • Delivers high purity DNA extraction for highly sensitive applications where sample is scarce
  • These beads provide an optimal surface for nucleic acid binding with high performance and low background
  • High magnetization (60 emu/g) and strong binding capacity giving fast magnetic response (∼5 secs) and shorten time required for magnetic steps during isolation
  • Beads come in two sizes (submicroscale diameter 700 nm and 400 nm), offering choice based on surface requirements and high binding capacity due to the small sized particles
  • Beads are monodispersed with narrow size distribution for consistent, reproducible results
  • Silanol hydroxyl groups on the bead surface give efficient, high purity isolation of nucleic acids
  • Tested for microbial contamination
  • Good buoyancy stability (low sedimentation rate): beads remain well suspended and dispersed for over 90 mins (at 1 mg/mL concentration)
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 20 mg/mL in water (0.05% sodium azide)
    Content And Storage Room Temperature
    Description Storage Solution: Water + 0.05% sodium azide
    Diameter (Metric) 400 nm
    For Use With (Application) Chaotropic salt chemistry and vortex mixing, roller mixing and sonication
    Product Type Silica coated, core-shell structure
    Quantity 60 mL
    Size 400 nm diameter particle
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.