Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERBP1 Monoclonal Antibody (OTI2C11), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA800719

Description
SERBP1 may play a role in the regulation of mRNA stability. It binds to the 3′-most 134 nt of the SERPINE1/PAI1 mRNA, a region which confers cyclic nucleotide regulation of message decay.Specifications
SERBP1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8NC51 | |
SERBP1 | |
Human recombit protein fragment corresponding to amino acids 1-262 of human SERBP1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2C11 | |
Unconjugated | |
SERBP1 | |
CGI-55, CHD3IP, HABP4L, PAI-RBP1, PAIRBP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
26135 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction