Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERBP1 Monoclonal Antibody (OTI2F6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526478
Description
SERBP1 Monoclonal Antibody for Western Blot, IHC (P)
SERBP1 may play a role in the regulation of mRNA stability. It binds to the 3'-most 134 nt of the SERPINE1/PAI1 mRNA, a region which confers cyclic nucleotide regulation of message decay.
Specifications
SERBP1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6AXS5, Q8NC51, Q9CY58 | |
SERBP1 | |
Human recombinant protein fragment corresponding to amino acids 1-262 of SERBP1 produced in E.coli. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2F6 | |
Unconjugated | |
SERBP1 | |
1200009K13Rik; 9330147J08Rik; AL022786; CGI-55; CHD3IP; chromodomain helicase DNA binding protein 3 interacting protein; HABP4L; hypothetical RNA binding protein RDA288; MGC128417 protein; PAI RBP 1; PAI-1 mRNA binding protein; PAI-1 mRNA-binding protein; PAI1 RNA-binding protein 1; Pairbp1; PAI-RBP1; plasminogen activator inhibitor 1 RNA binding protein; plasminogen activator inhibitor 1 RNA-binding protein; Rda288; RNA binding protein RDA288; SERBP1; Serpine1 mRNA binding protein 1; SERPINE1 mRNA-binding protein 1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
246303, 26135, 479535, 66870 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction