Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERBP1 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI2F6, lyophilized, TrueMAB™
SDP

Catalog No. 501665256
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-5256 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-5256 Supplier OriGene Supplier No. CF800689
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

SERBP1 may play a role in the regulation of mRNA stability. It binds to the 3′-most 134 nt of the SERPINE1/PAI1 mRNA, a region which confers cyclic nucleotide regulation of message decay.

Specifications

Antigen SERBP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2F6
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene SERBP1
Gene Accession No. Q6AXS5, Q8NC51, Q9CY58
Gene Alias CGI-55, CHD3IP, HABP4L, PAI-RBP1, PAIRBP1
Gene Symbols Serbp1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-262 of human SERBP1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246303, 26135, 479535, 66870
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.