Learn More
SERBP1 Mouse anti-Human, Clone: OTI2C11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800719

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SERBP1 may play a role in the regulation of mRNA stability. It binds to the 3′-most 134 nt of the SERPINE1/PAI1 mRNA, a region which confers cyclic nucleotide regulation of message decay.Specifications
SERBP1 | |
Monoclonal | |
Unconjugated | |
SERBP1 | |
CGI-55, CHD3IP, HABP4L, PAI-RBP1, PAIRBP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
26135 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2C11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8NC51 | |
SERBP1 | |
Human recombit protein fragment corresponding to amino acids 1-262 of human SERBP1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.