Learn More
Invitrogen™ SERBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113683
Description
Immunogen sequence: SYNYSDLDQS NVTEETPEGE EHHPVADTEN KENEVEEVKE EGPKEMTLDE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
SERBP1 may play a role in the regulation of mRNA stability. It binds to the 3'-most 134 nt of the SERPINE1/PAI1 mRNA, a region which confers cyclic nucleotide regulation of message decay.
Specifications
| SERBP1 | |
| Polyclonal | |
| Unconjugated | |
| SERBP1 | |
| 1200009K13Rik; 9330147J08Rik; AL022786; CGI-55; CHD3IP; chromodomain helicase DNA binding protein 3 interacting protein; HABP4L; hypothetical RNA binding protein RDA288; MGC128417 protein; PAI RBP 1; PAI-1 mRNA binding protein; PAI-1 mRNA-binding protein; PAI1 RNA-binding protein 1; Pairbp1; PAI-RBP1; plasminogen activator inhibitor 1 RNA binding protein; plasminogen activator inhibitor 1 RNA-binding protein; Rda288; RNA binding protein RDA288; SERBP1; Serpine1 mRNA binding protein 1; SERPINE1 mRNA-binding protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26135 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NC51 | |
| SERBP1 | |
| Synthetic peptide directed towards the middle region of human SERBP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.