Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115883
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115883 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115883 Supplier Invitrogen™ Supplier No. PA5115883
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SERBP1.

SERBP1 may play a role in the regulation of mRNA stability. It binds to the 3'-most 134 nt of the SERPINE1/PAI1 mRNA, a region which confers cyclic nucleotide regulation of message decay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERBP1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene SERBP1
Gene Accession No. Q6AXS5, Q8NC51, Q9CY58
Gene Alias 1200009K13Rik; 9330147J08Rik; AL022786; CGI-55; CHD3IP; chromodomain helicase DNA binding protein 3 interacting protein; HABP4L; hypothetical RNA binding protein RDA288; MGC128417 protein; PAI RBP 1; PAI-1 mRNA binding protein; PAI-1 mRNA-binding protein; PAI1 RNA-binding protein 1; Pairbp1; PAI-RBP1; plasminogen activator inhibitor 1 RNA binding protein; plasminogen activator inhibitor 1 RNA-binding protein; Rda288; RNA binding protein RDA288; SERBP1; Serpine1 mRNA binding protein 1; SERPINE1 mRNA-binding protein 1
Gene Symbols SERBP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SERBP1(Accession Q8NC51), corresponding to amino acid residues M288-S338.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246303, 26135, 66870
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.