Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599216
Description
Antibody detects endogenous levels of total SERC1.
SERINC1, also named as KIAA1253, TDE1L and TDE2, belongs to the TDE1 family. SERINC1 enhances the incorporation of serine into phosphatidylserine and sphingolipids.
Specifications
SERC1 | |
Polyclonal | |
Unconjugated | |
SERINC1 | |
1500011D18Rik; AI315070; Aigp2; AIGP-2; axotomy induced glycoprotein 2; axotomy-induced glyco/Golgi protein 2; KIAA1253; LRRGT00191; Membrane protein TMS-2; mKIAA1253; Serinc1; Serine incorporator 1; Tde1l; Tde2; TMS2; TMS-2; tumor differentially expressed 1, like; tumor differentially expressed 2; Tumor differentially expressed protein 1-like; Tumor differentially expressed protein 2; UNQ396/PRO732 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
294421, 56442, 57515 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q7TNK0, Q9NRX5, Q9QZI8 | |
SERINC1 | |
A synthesized peptide derived from human SERC1, corresponding to a region within C-terminal amino acids. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction