Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106534
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106534 Supplier Invitrogen™ Supplier No. PA5106534
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SERC1.

SERINC1, also named as KIAA1253, TDE1L and TDE2, belongs to the TDE1 family. SERINC1 enhances the incorporation of serine into phosphatidylserine and sphingolipids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERC1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SERINC1
Gene Accession No. Q7TNK0, Q9NRX5, Q9QZI8
Gene Alias 1500011D18Rik; AI315070; Aigp2; AIGP-2; axotomy induced glycoprotein 2; axotomy-induced glyco/Golgi protein 2; KIAA1253; LRRGT00191; Membrane protein TMS-2; mKIAA1253; Serinc1; Serine incorporator 1; Tde1l; Tde2; TMS2; TMS-2; tumor differentially expressed 1, like; tumor differentially expressed 2; Tumor differentially expressed protein 1-like; Tumor differentially expressed protein 2; UNQ396/PRO732
Gene Symbols SERINC1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SERC1, corresponding to a region within C-terminal amino acids.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294421, 56442, 57515
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.