Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101245
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101245 Supplier Invitrogen™ Supplier No. PA5101245
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SERINC2.

SERINC2 (Serine Incorporator 2) is a Protein Coding gene. Diseases associated with SERINC2 include Rapp-Hodgkin Syndrome and Alcohol Dependence. Among its related pathways are Viral mRNA Translation and Metabolism. Gene Ontology (GO) annotations related to this gene include L-serine transmembrane transporter activity. An important paralog of this gene is SERINC1. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERC2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SERINC2
Gene Accession No. Q8K0E7, Q96SA4
Gene Alias 2310004K20Rik; AW121759; FKSG84; PRO0899; Serinc2; Serine incorporator 2; TDE2; TDE2L; tumor differentially expressed protein 2; Tumor differentially expressed protein 2-like; UNQ263/PRO300
Gene Symbols SERINC2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SERINC2(Accession Q96SA4), corresponding to amino acid residues A271-A321.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230779, 313057, 347735
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.