Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERCA1 ATPase Antibody (SR1065), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | SERCA1 ATPase |
---|---|
Clone | SR1065 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
SERCA1 ATPase Monoclonal antibody specifically detects SERCA1 ATPase in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
SERCA1 ATPase | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ATP2A, ATPase, Ca++ transporting, cardiac muscle, fast twitch 1, Calcium pump 1, Calcium-transporting ATPase sarcoplasmic reticulum type, fast twitch skeletalmuscle isoform, EC 3.6.3, Endoplasmic reticulum class 1/2 Ca(2+) ATPase, sarcoplasmic/endoplasmic reticulum calcium ATPase 1, SERCA1EC 3.6.3.8, SR Ca(2+)-ATPase 1 | |
A synthesized peptide derived from human SERCA1 ATPase (Uniprot #: O14983) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1065 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
487 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title