Learn More
Invitrogen™ SERCA1 ATPase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587633
Description
Positive Samples: HL-60; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein, Sarcoplasmic reticulum membrane Immunogen sequence: LSVLVTIEMC NALNSLSENQ SLLRMPPWVN IWLLGSICLS MSLHFLILYV DPLPMIFKLR ALDLTQWLMV LKISLPVIGL DEILKFVARN YLEDPEDERR K.
ATP dependent calcium pumps are responsible, in part, for the maintenance of low cytoplasmic free calcium concentrations. The ATP pumps that reside in intracellular organelles are encoded by a family of structurally related enzymes, termed the sarcoplasmic or endoplasmic reticulum calcium (SERCA) ATPases. The SERCA1 gene is exclusively expressed in type II (fast) skeletal muscle. The SERCA2 gene is subject to tissue dependent processing which is responsible for the generation of SERCA2a muscle-specific form expressed in type I (slow) skeletal, cardiac and smooth muscle and the SERCA2b isoform expressed in all cell types. The SERCA3 gene is not as well characterized and is found in non-muscle cells.
Specifications
SERCA1 ATPase | |
Polyclonal | |
Unconjugated | |
ATP2A1 | |
adult Ca2+ ATPase; ATP2A; ATP2A1; ATP2A3; ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 1; ATPase, Ca++ transporting, cardiac muscle, fast twitch 1; ATPase, Ca++ transporting, ubiquitous; Ca2+ ATPase; calcium pump 1; Calcium-transporting ATPase sarcoplasmic reticulum type, fast twitch skeletal muscle isoform; endoplasmic reticulum class 1/2 Ca(2+) ATPase; neonatal Ca2+ ATPase; sarcoplasmic/endoplasmic reticulum calcium ATPase 1; SERCA; SERCA ATPase; Serca1; SERCA1a; SR Ca(2+)-ATPase 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
11937, 487 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14983, Q8R429 | |
ATP2A1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 901-1001 of human SERCA1/SERCA1/ATP2A1 (NP_7752931). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.