Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERCA2 ATPase Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100237UV
Description
SERCA2 ATPase Polyclonal antibody specifically detects SERCA2 ATPase in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
SERCA2 ATPase | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
ATP2BFLJ20293, ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2, ATPase, Ca++ transporting, cardiac muscle, slow twitch 2, Calcium pump 2, Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletalmuscle isoform, cardiac Ca2+ ATPase, DAR, DD, EC 3.6.3, EC 3.6.3.8, Endoplasmic reticulum class 1/2 Ca(2+) ATPase, FLJ38063, MGC45367, sarcoplasmic/endoplasmic reticulum calcium ATPase 2, SERCA2DKFZp686P0211, SR Ca(2+)-ATPase 2 | |
The immunogen recognized by this antibody maps to a region between residue 475 and 525 of human ATPase, Ca++ transporter, cardiac muscle, slow twitch 2 using the numbering given in entry NP_733765.1 (GeneID 488). | |
0.1 mL | |
Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
488 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction