Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERCA2 ATPase Antibody (IID8), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB300529C
Description
SERCA2 ATPase Monoclonal specifically detects SERCA2 ATPase in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| SERCA2 ATPase | |
| Monoclonal | |
| DyLight 650 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ATP2A2 | |
| Purified canine cardiac sarcoplasmic reticulum. | |
| Primary | |
| Detects Sarcoplasmic or Endoplasmic Reticulum Calcium 2 (SERCA2) ATPase. This does not discriminate between the two isoforms. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| IID8 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| ATP2BFLJ20293, ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2, ATPase, Ca++ transporting, cardiac muscle, slow twitch 2, Calcium pump 2, Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletalmuscle isoform, cardiac Ca2+ ATPase, DAR, DD, EC 3.6.3, EC 3.6.3.8, Endoplasmic reticulum class 1/2 Ca(2+) ATPase, FLJ38063, MGC45367, sarcoplasmic/endoplasmic reticulum calcium ATPase 2, SERCA2DKFZp686P0211, SR Ca(2+)-ATPase 2 | |
| Mouse | |
| 0.1 mL | |
| 488 | |
| Human, Mouse, Rat, Pig, Amphibian, Bovine, Canine, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Virus | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction