Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERCA2 ATPase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142877
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142877 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142877 Supplier Invitrogen™ Supplier No. PA5142877
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize both reported isoforms (NP_001672.1; NP_733765.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

This gene encodes one of the SERCA Ca(2+)-ATPases, which are intracellular pumps located in the sarcoplasmic or endoplasmic reticula of muscle cells. This enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol into the sarcoplasmic reticulum lumen, and is involved in regulation of the contraction/relaxation cycle. Mutations in this gene cause Darier-White disease, also known as keratosis follicularis, an autosomal dominant skin disorder characterized by loss of adhesion between epidermal cells and abnormal keratinization. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERCA2 ATPase
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Atp2a2
Gene Accession No. O55143, P11507, P16615
Gene Alias 9530097L16Rik; ATP2; Atp2a2; ATP2B; ATPase Ca++ transporting cardiac muscle slow twitch 2; ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 2; ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2; ATPase, Ca++ transporting, cardiac muscle, slow twitch 2; ATPase, Ca++ transporting, slow twitch 2; Ca(2+)-transport ATPase class 3; calcium ATPase; calcium pump 2; calcium-ATPase (EC 3.6.1.3); calcium-transporting ATPase; calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletal muscle isoform; cardiac Ca2+ ATPase; D5Wsu150e; DAR; DD; endoplasmic reticulum class 1/2 Ca(2+) ATPase; mKIAA4195; putative SERCA isoform; sarco(endo)plasmic reticulum Ca(2+)-dependent ATPase 2; sarco/endoplasmic reticulum Ca2+-ATPase 2; sarcoplasmic reticulum Ca2+-transport ATPase isoform; sarcoplasmic/endoplasmic reticulum calcium ATPase 2; sarcoplasmic/endoplasmic-reticulum Ca(2+) pump gene 2; SERCA; SERCA ATPase; SERCA2; Serca2a; SERCA2B; SercaII; SR Ca(2+)-ATPase 2; unnamed protein product
Gene Symbols Atp2a2
Host Species Goat
Immunogen Peptide with sequence DELNPSAQRDACLN.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11938, 29693, 488
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.