Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERCA3 ATPase Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578838
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Skeletal Muscle Tissue, Mouse Skeletal Muscle Tissue. IHC: Mouse Thymus Tissue, Rat Thymus Tissue.
ATP dependent calcium pumps are responsible in part for the maintenance of low cytoplasmic free calcium concentrations. The ATP pumps that reside in intracellular organelles are encoded by a family of structurally related enzymes, termed the sarcoplasmic or endoplasmic reticulum calcium ATPases (SERCA). The SERCA2 gene is subject to tissue dependent processing which is responsible for the generation of SERCA2a muscle-specific isoform expressed in type I (slow) skeletal, cardiac and smooth muscle and the SERCA2b isoform expressed in all cell types. SERCA3 has been found to co-express with SERCA2b in platelets, mast cells, lymphoid cells, and epithelial cells. Due to this pattern of expression it is referred to as the non-muscle form of SERCA.
Specifications
SERCA3 ATPase | |
Polyclonal | |
Unconjugated | |
Atp2a3 | |
adenosine triphosphatase, calcium; ATP2A3; ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 3; ATPase, Ca(2+)-transporting, ubiquitous; ATPase, Ca++ transporting, ubiquitous; calcium pump 3; calcium-translocating P-type ATPase; sarco/endoplasmic reticulum Ca2+ -ATPase; sarcoendoplasmic reticulum Ca2+ ATPase type 3; sarcoplasmic/endoplasmic reticulum calcium ATPase 3; SERCA3; SERCA3b; SR Ca(2+)-ATPase 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25391, 489, 53313 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
P18596, Q64518, Q93084 | |
Atp2a3 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human ATP2A3 (1-30aa MEAAHLLPAADVLRHFSVTAEGGLSPAQVT). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction