Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERF1A Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178392AF750
Description
SERF1A Polyclonal specifically detects SERF1A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SERF1A | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
FAM2B, h4F5, H4F5small EDRK-rich factor 1,4F5, Protein 4F5, SERF1, SMA modifier 1, small EDRK-rich factor 1A (telomeric), SMAM1FAM2ASERF1B, spinal muscular atrophy-related gene H4F5 | |
Rabbit | |
0.1 mL | |
8293 | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
SERF1A | |
A synthetic peptide made to the N-terminal portion of the human SERF1A protein (between residues 1-60) [UniProt O75920] | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction