Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERF1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567379
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567379 Supplier Invitrogen™ Supplier No. PA567379
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSGGQKSESKM SAGPHLPLKA PRENPCFPLP AAGGSR Highest antigen sequence identity to the following orthologs - mouse 38%, rat 38%.

This gene is part of a 500 kb inverted duplication on chromosome 5q13. This duplicated region contains at least four genes and repetitive elements which make it prone to rearrangements and deletions. The repetitiveness and complexity of the sequence have also caused difficulty in determining the organization of this genomic region. The duplication region includes both a telomeric and a centromeric copy of this gene. Deletions of this gene, the telomeric copy, often accompany deletions of the neighboring SMN1 gene in spinal muscular atrophy (SMA) patients, and so it is thought that this gene may be a modifier of the SMA phenotype. The function of this protein is not known; however, it bears low-level homology with the RNA-binding domain of matrin-cyclophilin, a protein which colocalizes with small nuclear ribonucleoproteins (snRNPs) and the SMN1 gene product. Alternatively spliced transcripts have been documented but it is unclear whether alternative splicing occurs for both the centromeric and telomeric copies of the gene. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERF1A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SERF1A
Gene Accession No. O75920
Gene Alias 4F5; FAM2A; FAM2B; h4F5; protein 4F5; SERF1; SERF1A; SERF1B; SMA modifier 1; small EDRK-rich factor 1; small EDRK-rich factor 1A; small EDRK-rich factor 1A (telomeric); small EDRK-rich factor 1B (centromeric); SMAM1; spinal muscular atrophy-related gene H4F5; Unknown (protein for MGC:133508)
Gene Symbols SERF1A
Host Species Rabbit
Immunogen Recombinant Human SERF1A. Recombinant protein control fragment (Product #RP-105668).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8293
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.