Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERGEF Monoclonal Antibody (1A2)
GREENER_CHOICE

Catalog No. PIMA521645
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521645 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521645 Supplier Invitrogen™ Supplier No. MA521645
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: KHGQLANEAA FLPVPQKIEA HCFQNEKVTA IWSGWTHLVA QTETGKMFTW GRADYGQLGR KLETYEGWKL EKQDSFLPCS RPPNSMPSSP HCLTGATE.

Probable guanine nucleotide exchange factor (GEF), which may be involved in the secretion process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERGEF
Applications ELISA
Classification Monoclonal
Clone 1A2
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SERGEF
Gene Accession No. Q9UGK8
Gene Alias deafness locus associated putative guanine nucleotide exchange factor; deafness locus-associated putative guanine nucleotide exchange factor; DelGEF; Gef; Gnef; Gnefr; guanine nucleotide exchange factor (RCC1 related); guanine nucleotide exchange factor-related protein; secretion regulating guanine nucleotide exchange factor; secretion-regulating guanine nucleotide exchange factor; Sergef
Gene Symbols SERGEF
Host Species Mouse
Immunogen SERGEF (NP_036271, 240 a.a. ~ 337 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26297
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.