Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERGEF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550124
Description
The antibody detects endogenous levels of total SERGEF protein.
Probable guanine nucleotide exchange factor (GEF), which may be involved in the secretion process.
Specifications
SERGEF | |
Polyclonal | |
Unconjugated | |
SERGEF | |
deafness locus associated putative guanine nucleotide exchange factor; deafness locus-associated putative guanine nucleotide exchange factor; DelGEF; Gef; Gnef; Gnefr; guanine nucleotide exchange factor (RCC1 related); guanine nucleotide exchange factor-related protein; secretion regulating guanine nucleotide exchange factor; secretion-regulating guanine nucleotide exchange factor; Sergef | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26297, 27414 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q80YD6, Q9UGK8 | |
SERGEF | |
Synthesized peptide derived from internal of human SERGEF (aa 179-228). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction