Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERGEF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106727
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106727 Supplier Thermo Scientific Supplier No. PA5106727
Add to Cart
Add to Cart

SERGEF Polyclonal Antibody

SERGEF Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen SERGEF
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SERGEF
Gene Accession No. Q80YD6, Q9UGK8
Gene Alias deafness locus associated putative guanine nucleotide exchange factor; deafness locus-associated putative guanine nucleotide exchange factor; DelGEF; Gef; Gnef; Gnefr; guanine nucleotide exchange factor (RCC1 related); guanine nucleotide exchange factor-related protein; secretion regulating guanine nucleotide exchange factor; secretion-regulating guanine nucleotide exchange factor; Sergef
Gene Symbols SERGEF
Host Species Rabbit
Immunogen A synthesized peptide derived from human SERGEF(Accession Q9UGK8), corresponding to amino acid residues G178-T228.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26297, 27414
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.