Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Serglycin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550794 Supplier Invitrogen™ Supplier No. PA550794
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total Serglycin protein.

This gene encodes a protein best known as a hematopoietic cell granule proteoglycan. Proteoglycans stored in the secretory granules of many hematopoietic cells also contain a protease-resistant peptide core, which may be important for neutralizing hydrolytic enzymes. This encoded protein was found to be associated with the macromolecular complex of granzymes and perforin, which may serve as a mediator of granule-mediated apoptosis. Two transcript variants, only one of them protein-coding, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serglycin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SRGN
Gene Accession No. P10124
Gene Alias gp600; hematopoetic proteoglycan core peptide; hematopoetic proteoglycan core protein; hematopoietic proteoglycan core protein; Mastocytoma proteoglycan core protein; p.PG; Platelet proteoglycan core protein; PPG; Prg; Prg1; proteoglycan 1, secretory granule; proteoglycan protein core for mast cell secretory granule; proteoglycan, secretory granule; secretory granule proteoglycan core peptide; secretory granule proteoglycan core protein; Serglycin; serglycin proteoglycan; Sgc; Srgn
Gene Symbols SRGN
Host Species Rabbit
Immunogen Full length fusion protein of Human SRGN.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5552
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.