Learn More
Invitrogen™ Serglycin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113691
Description
Immunogen sequence: LLKCSRLVLA LALILVLESS VQGYPTRRAR YQWVRCNPDS NSANCLEEKG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%; Mouse: 91%; Rabbit: 85%; Rat: 91%.
This gene encodes a protein best known as a hematopoietic cell granule proteoglycan. Proteoglycans stored in the secretory granules of many hematopoietic cells also contain a protease-resistant peptide core, which may be important for neutralizing hydrolytic enzymes. This encoded protein was found to be associated with the macromolecular complex of granzymes and perforin, which may serve as a mediator of granule-mediated apoptosis. Two transcript variants, only one of them protein-coding, have been found for this gene.
Specifications
| Serglycin | |
| Polyclonal | |
| Unconjugated | |
| SRGN | |
| gp600; hematopoetic proteoglycan core peptide; hematopoetic proteoglycan core protein; hematopoietic proteoglycan core protein; Mastocytoma proteoglycan core protein; p.PG; Platelet proteoglycan core protein; PPG; Prg; Prg1; proteoglycan 1, secretory granule; proteoglycan protein core for mast cell secretory granule; proteoglycan, secretory granule; secretory granule proteoglycan core peptide; secretory granule proteoglycan core protein; Serglycin; serglycin proteoglycan; Sgc; Srgn | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5552 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P10124 | |
| SRGN | |
| Immunogen sequence: LLKCSRLVLA LALILVLESS VQGYPTRRAR YQWVRCNPDS NSANCLEEKG (aa 5-54). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.