Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERHL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570408
Description
This target displays homology in the following species: Human: 100%; Mouse: 79%; Rabbit: 79%; Rat: 93%.
SERHL2 is a protein coding gene.
Specifications
SERHL2 | |
Polyclonal | |
Unconjugated | |
SERHL2 | |
dJ222E13.1; RGD1563386; SERHL; Serhl2; serine hydrolase like 2; serine hydrolase-like 2; serine hydrolase-like protein 2; testis secretory sperm-binding protein Li 216e | |
Rabbit | |
Affinity chromatography | |
RUO | |
253190 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H4I8 | |
SERHL2 | |
synthetic peptide directed towards the C-terminal region of human SERHL2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction