Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Series 210 Laser Biopsy Cassettes with Front Tab Lids
Shop All Epredia Products
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lilac
Ochre
Orange
Pink
Red
White
Yellow
Description
- Epredia™ Laser Cassettes are designed for use on both the Epredia™ NOVA™ and VEGA™ Laser Printers
- Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
- Comes in multiple different colors with color-matched lids
- Designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
- Comes in taped stacks of 40 cassettes; 25 stacks per case for a total of 1000 cassettes with separate lids
- Not compatible with thermal tape style printers
Specifications
Specifications
Pore Style | Small Squares |
Closure Included | Yes |
Quantity | 40 Cassettes/Taped Stack, 25 Stacks/Cs. |
Closure Type | Separate Lid with Tab |
Color | Red |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction