Learn More
Epredia™ Series 215 Laser Biopsy Cassettes with Lids
Catalog No. 2151110LM Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lilac
Ochre
Orange
Pink
Red
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-511-10LM Ochre
2150110LM White
2150210LM Yellow
2150310LM Blue
2150410LM Pink
2150510LM Green
2150610LM Orange
2150710LM Gray
2150810LM Aqua
2150910LM Lilac
2151210LM Red
21-510-10LM Beige
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 21-511-10LM Supplier Epredia™ Supplier No. 2151110LM
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Manufactured specifically for laser printing, the Epredia™ Series 215 Biopsy Cassettes come with separate lids and are designed for small tissue samples.
  • Epredia™ Laser Cassettes are designed for use on both the NOVA™ and VEGA™ Laser Printers.
  • They contain a special constituent chemical compound that carbonizes when exposed to the laser beam.
  • Each style comes in 10 different colors: white, yellow, blue, pink, green, orange, grey, aqua, lilac and red with color-matched lids.
  • Each cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing.
  • Series 110, 115 and 215 come in taped stacks of 40 cassettes; 25 stacks per case.
  • Series 815 come in taped stacks of 40 cassettes with lids attached; 45 stacks per case.
  • The Epredia™ Laser Cassette is not compatible with thermal tape style printers.
Specifications
Pore Style Small Squares
Quantity 40 Cassettes/Taped Stack, 25 Stacks/Cs.
Closure Included Yes
Closure Type Separate Lid
Color Ochre
Product Type Cassette