Learn More
Johannes Lieder Series for Secondary Schools, Set No. III. Organs of Sense
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Description
- 4450 Set No. III. Organs of Sense.
- 16 Microscope Slides
- Paramaecium, silvered to show the neuroformative system
- Lumbricus, earthworm, t.s. with ventral nerve cord
- Insect brain, frontal l.s.
- Planaria, sec. through ocelli
- Haliotis, marine snail, pinhole camera eye l.s.
- Helix, snail, eye l.s.
- Alloteuthis, cuttlefish, camera eye l.s.
- Compound eye of an insect, l.s.
- Young rat, head with eyes t.s.
- Retina of cat, t.s. showing rods and cones
- Internal ear (cochlea) from guinea pig, l.s.
- Taste buds from tongue of rabbit, t.s.
- Peripheral nerve fibres, osmic acid material showing Ranvier's nodes
- Spinal cord of cat t.s. with large motor nerve cells
- Cerebellum of cat, t.s. routine stained
- Cerebrum of cat, t.s. silvered to show the pyramid cells
Specifications
Specifications
Classification | Histology |
Material | Elastin Stained Glass |
Mounting Type | Cross and Longitudinal Section |
Type | Microscope Slides |
Slide Type | Prepared Slide |