Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serine carboxypeptidase 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987817
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7817 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7817 Supplier Bioss Supplier No. BS7542R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be involved in vascular wall and kidney homeostasis. Target protein may act as a serine carboxypeptidase.

Specifications

Antigen Serine carboxypeptidase 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SCPEP1
Gene Accession No. Q9HB40
Gene Alias 2410018F01Rik; 4833411K15Rik; AI957256; HSCP1; MSTP034; retinoid inducible serine carboxypeptidase; Retinoid-inducible serine carboxypeptidase; retinoid-inducible serine caroboxypeptidase; retinoid-inducible serine caroboxypetidase; Risc; SCP1; SCPEP1; Serine carboxypeptidase 1; serine carboxypeptidase 1 precursor protein; serine caroboxypeptidase 1; UNQ265/PRO302
Gene Symbols SCPEP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RISC/Serine carboxypeptidase 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59342
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.