Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serine protease 23 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306049PE
Description
Serine protease 23 Polyclonal antibody specifically detects Serine protease 23 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Serine protease 23 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, MGC5107, protease, serine, 23, Putative secreted protein Zsig13, serine protease 23, serine protease, umbilical endothelium, SIG13, SPUVE, ZSIG13 | |
Produced in rabbits immunized with E. coli-derived Human Serine protease 23 fragment. | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
11098 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction